Sodium ethoxide 21% in ethanol, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Sodium ethylate, Sodium ethanolate
CAS Number:141-52-6
:
Formula:C₂H₅NaO
MDL Number:MFCD00012417
Boiling Pt:91 °C
Melting Pt:260
Density:0.868 g/cm³
Molecular Weight:68.05 g/mol
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:205-487-5
UN:2924
ADR:3 II
:GHS05
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow to brown Solution (May darken on storage)
- Titration with HCl:20.0 to 22.0 %
- NaOH + Na2CO3 (as NaOH):≤0.5 %
Specification Test Results
Frequently Bought Together