4-Fluoro-2-methylbenzenesulphonyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-Fluoro-2-methylbenzene-1-sulphonyl chloride
CAS Number:7079-48-3
:
Formula:C₇H₆ClFO₂S
MDL Number:MFCD03094390
Boiling Pt:245…246 °C
Density:1.433
Molecular Weight:208.64 g/mol
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:3265
ADR:8 II
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together