Order Entry
Slovenia
ContactUsLinkComponent
Naphthol AS-D chloroacetate, Sigma-Aldrich®
Naphthol AS-D chloroacetate, Sigma-Aldrich®
Catalog # 905-10CAP-S
CAS Number:  
Naphthol AS-D chloroacetate, Sigma-Aldrich®
Catalog # 905-10CAP-S
CAS Number:  

Specifications

  • Pk:
    10
  • Seq:
    0010
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance (colour):
    White
  • Appearance (form):
    Crystalline Powder
  • Performance:
    Conforms

Specification Test Results

About this item

Synonyms: 3-[[(2-Methylphenyl)amino]carbonyl]-2-naphthyl chloroacetate
 :
 :353.8 g/mol
EINECS:252-463-5
 :1
CAS Number:35245-26-2
Formula:C₂₀H₁₆ClNO₃
 :M-Clarity™ quality level = MQ500