(1S)-(+)-Camphor-10-sulphonic acid, (max. 2% H₂O) 98+% (dry weight)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: ((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, (1S)-(+)-2-Oxobornane-10-sulphonic acid, D(+)-Camphor-10-sulphonic acid, (1S)-(+)-7,7-Dimethylbicyclo(2.2.1)-2-heptanon-1-methane sulphonic acid, (1S)-(+)-10-Camphorsulphonic acid, D-Camphorsulphonic acid
CAS Number:3144-16-9
:
Formula:C₁₀H₁₆O₄S
MDL Number:MFCD00064157
Boiling Pt:375
Molecular Weight:232.3 g/mol
Melting Pt:196…200 °C (dec.)
ADR:8 III
Merck Index:14,1734
EINECS:221-554-1
UN:3261
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together