Acetonitrile ≥99.9%, BAKER ANALYZED® HPLC, Ultra Gradient for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
Boiling Pt:81.6
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
Molecular Weight:41.05 g/mol
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
UN:1648
ADR:3 II
Merck Index:13,00071
EINECS:200-835-2
REACH:01-2119471307-38
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (by GC):min. 99.9 %
- Appearance:passes test
- Distilling Range 80-82°C:min. 95%
- Identity (by IR):passes test
- Transmittance 240 nm - 420 nm:min. 98 %
- Water (H2O):max. 0.01 %
- Measured at Emission Maximum for Solvent Impurities:max. 1.0
- Measured at 450 nm:max. 0.3
- at 254 nm:max. 0.0005
- Fluorescence (in ppb, measured as Benzo(a)Pyrene at EX/Em=280nm/Total Emission:max. 0.5
- at 210 nm:max. 0.002
- Residue after Evaporation:max. 1 ppm
- Titrable Acid (meq/g):max. 0.0008
- Titrable Base (meq/g):max. 0.0006
- at 200 nm:max. 0.05
- at 220 nm:max. 0.01
- at 254-400 nm:max. 0.01
- UV Cut-off, nm:max. 190
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_BAKR9017
SDS
VWR J.T.Baker® High Purity solvents and reagents
Product Literature
Frequently Bought Together