Lithium hydroxide monohydrate (∼56% LiOH), extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1310-66-3
:
Formula:LiOH·1H₂O
MDL Number:MFCD00149772
Boiling Pt:920 °C (760 mmHg)
Molecular Weight:41.96 g/mol
Melting Pt:462 °C
ADR:8 II
Merck Index:13,05556
EINECS:215-183-4
UN:2680
:GHS07
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to cream to yellow Crystalline powder
- Titration with HCl:≥54 % (LiOH)
- Chloride (Cl):≤500 ppm
- Sulfate (SO4):≤500 ppm
- Aluminium (Al):≤300 ppm
- Iron (Fe):≤10 ppm
- Sodium + Potassium (Na+K):≤3000 ppm
- Carbon dioxide (CO2):≤3500 ppm
- Insoluble matter:≤0.1 % (in water)
Specification Test Results
Frequently Bought Together