Hydrogen peroxide 50% (w/w) in water stabilised, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Boiling Pt:114 °C (760 mmHg)
Melting Pt:–52 °C
Density:1.2 g/cm³
Molecular Weight:34.01 g/mol
Storage Temperature:Refrigerator
EINECS:231-765-0
ADR:5.1 II
Merck Index:14,04798
UN:2014
:GHS07
:GHS05
:GHS03
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Solution
- Refractive index:1.3665 to 1.3685 (20°C, 589 nm)
- Density:(20°C) 1.19 to 1.21 kg/l
- Stabilizer:≤200 ppm phosphates
- pH:1 to 4
- Titration with KMnO4:49.8 to 50.6 %
Specification Test Results
Frequently Bought Together