Automated crude fibre determination, FIBRETHERM®
Catalog # 705-0582
Supplier: GERHARDT
Specifications
- Description:Automated crude fibre determination, FIBRETHERM®
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Automated processing of the boiling and filtration steps for the crude fibre, ADF and NDF determination. Ideal for laboratories with high sample throughput.
- Saves time with simultaneous and reproducible analysis of up to 12 samples
- Small footprint and ergonomic design, including pneumatic lift and errors alarm
- Automation of procedure reduces costs and reagents used
- FibreBags provide constant filtration quality
- Safe operation with additions of water, acid and caustic reagents and subsequent filtration that can be programmed
Delivery: System is delivered complete.