VWR® MiniStar silverline, Microcentrifuge
This centrifuge was developed for applications that do not require high speed centrifugation, such as microfiltration and cell separation, and can also be used in a cold room. On closing the lid, the rotor quickly accelerates to maximum speed. Pressing the button on the front of the unit stops operation and opens the lid. Alternatively, the unit can be turned on and off with the switch at the back.
- Metal base with ABS body and transparent safety cover
- Quiet and compact design
- Starts and stops in seconds
- Easy to operate, for eight microtubes or four strip tubes
: MiniStar silverline supplied with 8×1,5/2,0 ml rotor, 4×(8×0,2 ml) strip tube rotor, eight adapters for 0,5 ml and eight adapters for 0,4 ml tubes.
Frequently Bought Together