Order Entry
Slovenia
ContactUsLinkComponent
Reader, Micro-Haematocrit
Reader, Micro-Haematocrit
Catalog # 720-0100
Supplier:  HAWKSLEY AND SONS
undefined
Reader, Micro-Haematocrit
Catalog # 720-0100
Supplier:  HAWKSLEY AND SONS
Specifications
  • Description:
    Reader, Micro-Haematocrit, column height: 40-70 mm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
For determination of packed cell volume, using 75 mm capillary tubes with total fluid column heights between 40 and 70 mm.