Sample collection systems, Samplit™, SP Bel-Art
231-0012
- Type:Sterile
- Capacity:190 ml
- Description:Sample jar with scoop
- Seq:01
- Environmentally Preferable:
- Pk:25
PS. This sample collection system comprises a sample jar with integral scoop, allowing samples to be transferred easily and directly to a collection receptacle. Ideally suited for bulk materials, solid, semi-solid, and other materals.
- Available in sterile or non sterile versions
- Designed for rough handling
- Semicircular, extremely robust scoop with peripheral cutting edge