Glass capillaries and graduated micropipettes, disposable
: CAMAG
Disposable glass capillaries for manual sample application of 0,5, 1, 2, 5 or 10 µl. They can be handheld and positioned with the Multipurpose Spotting Guide (551-4230). They can also be used with Universal Capillary Holder (552-0044). Graduated 5 µl glass micropipettes are suitable for qualitative analysis on conventional TLC layers.
- Colour coded vials
- Convenient and user friendly
- Micropipettes are graduated in microlitres
Frequently Bought Together