White microplates for luminescence applications
Supplier: Thermo Fisher Scientific
Certificates
About this item
Microlite™ microplates are densely pigmented to minimise crosstalk and to provide high reflectivity. These characteristics are ideal for intense luminescent reactions where light piping may be a concern or minimal luminescence assays where even a minute amount of light could affect results.
- Microlite™+ surface offers minimum crosstalk and extra high reflectivity
- Choice of 96-well or strip plate format
- Flat bottom wells for optimum optical characteristics
Microlite™ 1+ is the ideal vessel for low signal luminescent reactions, offering medium binding with enhanced dynamic range, extra high reflectivity and minimal crosstalk.
Microlite™ 2+ is an irradiated, high binding surface with enhanced dynamic range, extra high reflectivity and minimal crosstalk for low signal luminescent reactions.
Specifications
- Catalog No:
- 735-0525
- 735-0526
- 735-0524
- 735-0523
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement