Tangential flow filtration capsules, Minimate™
Supplier: Cytiva (Formerly Pall Lab)
Certificates
About this item
The disposable Minimate™ TFF capsules, with Omega™ membrane, can perform concentration processes desalting or buffer exchange on the same system with minimal user intervention, and achieves high concentration factors in a single step due to the low hold-up volume of the devices.
- Scaleable
- Works with a variety of positive pressure laboratory pumps
- High product recovery
- 100% integrity tested
The capsules will process samples of up to 1 litre, or efficiently concentrate samples as little as 5 ml.
Delivery: Supplied with all fittings and tubing to simplify installation into filtration setups. A Certificate of Quality is included with each capsule.
Frequently Bought Together