Petri dishes, Sterilin®
Supplier: Sterilin
Certificates
About this item
Ideal for use when savings in media or incubator space are required.
- Aseptically manufactured under cleanroom conditions (Class 7 ISO 14644) to exclude microbiological contamination
- 55 mm dish accommodates 47 mm membrane filters making it suitable for water testing
- 50 mm deep form dish is over 20 mm deep and designed for use with liquid media
Dimensions shown are for outer Ø base and outer height (mm).
Save incubator space with Thermo Scientific™ Sterilin™ 30 to 140 mm Petri Dishes. These small dishes are suitable for most applications. Not available in America.
140 mm dish is produced and tested in accordance with the EN ISO 24998 Petri dish standard which includes stringent dimensional controls.
Frequently Bought Together