VWR®, Face Masks, 3 ply, Non Woven PP
Face masks made from 3 ply, non woven PP featuring a nose bridge and ear loops or ties.
- Latex-free
- Bacterial filtration efficiency: 90%
- Particle filtration efficiency: 98,5%
- Particle size: 0,1 μm
- For industrial and cleanroom use only; do not use for protection against COVID-19
- This product is not a medical device for medical or clinical use within the meaning of Regulation (EU) 2017/745/Directive 93/42/EEC
- This product is not a personal protective equipment (PPE) within the meaning of Regulation (EU) 2016/425
Density: 20 g/m² (front layer); 25 g/m² (back layer); 20 g/m² (centre).
CoC VWR Face Masks, 3 Ply, Non Woven PP COC_VWRI113-0714_D9210085 COC_VWRI113-0714_D9211143 COC_VWRI113-0716_D210097 COC_VWRI113-0716_D9200115
Frequently Bought Together