Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Wash bottles, glass
Wash bottles, glass
Wash bottle complete, Wash bottle complete
Catalog # 215-6290
Supplier:   Lenz Laborglas
Wash bottles, glass
Catalog # 215-6290
Supplier:  Lenz Laborglas
Wash bottles, glass
Wash bottles, glass
Wash bottle complete, Wash bottle complete
Price
66,00 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass clear.
  • Very good chemical resistance
  • Very good thermal resistance
  • Fully interchangeable parts
Universal wash bottle consisting of a 1000 ml capacity flat bottomed flask with standard ground joint sleeve NS 29/32 and spray assembly with standard ground joint cone NS 29/32.
Documents
Specifications
  • Type:
    Wash bottle complete
  • Description:
    Wash bottle complete
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Spare wash head with joint NS 29/32 for 1000 ml bottle
    Spare wash head with joint NS 29/32 for 1000 ml bottle
    215-6292
    Log in for availability
    1 * 1 items - 26,80 €