Work with ease and filter with confidence with Stericup® and Steritop® filtration systems. The Stericup® E and Steritop® E sterile filters are designed as a greener alternative for vacuum sterile filtration that significantly reduces plastic and packaging.
- The filter collar disconnects from the receiver bottle with just a quarter turn reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood
- Bold product identification on the filter collar plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection
- Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilised contents
- Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears
- Click seal cap rests on the side to avoid risk of contamination
- Receiver bottles are stackable to save storage space
This Steritop® E is a 38 mm thread collar which uses a 0,22 μm pore size polyethersulfone (PES) membrane filter unit. The 38 mm thread directly connects to our media bottles and majority of other standard commercial media bottles. Use a 45 mm threaded glass or plastic receiver bottle no larger than 2 liters.
This Steritop® E is a 45 mm thread collar which uses a 0,22 μm pore size polyethersulfone (PES) membrane filter unit. The 45 mm thread directly connects to wider neck media bottles (such as Gibco® or glass bottles). Use a 45 mm threaded glass or plastic receiver bottle no larger than 2 liters.