Hypoxia Incubator Chamber
Catalog # STMC27310
Supplier: STEMCELL Technologies
Specifications
- Description:Hypoxia incubator chamber
- Environmentally Preferable:
- Pk:1 KIT
Specifications
About this item
The Hypoxia incubator chamber is for generation of a hypoxic environment for tissue culture. This is a self-contained and sealed chamber that fits inside existing laboratory incubators.
- All units are molded from high quality polycarbonate and will not break, crack or cloud
- Integrated stacking feature for storage
- Guaranteed for 1 year against defects, leaks and breakage
Each unit utilizes a surface-type seal in which all portions of the O-ring are uniformly compressed by a ring clamp for a reliable, air-tight seal. The cylindrical walls and semi-spherical top and bottom minimise gas flow resistance and eliminate dead space during initial purging.
This chamber can use with 84×35 mm plates, 27×60 mm plates, 12×100 mm plates, 12×96-well plates and 18×25 cm² flasks.
Ordering information: Includes with polycarbonate base with O-ring and tubing clamp, polycarbonate lid and tray, ring clamp, O-ring and tubing clamp with tubing.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').