Order Entry
Slovenia
ContactUsLinkComponent
Anti-Calprotectin Mouse Monoclonal Antibody [clone: CPT/1028]
Anti-Calprotectin Mouse Monoclonal Antibody [clone: CPT/1028]
Catalog # ANTIA251059-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Calprotectin Mouse Monoclonal Antibody [clone: CPT/1028]
Catalog # ANTIA251059-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Calprotectin
  • Clonality:
    Monoclonal
  • Clone:
    CPT/1028
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgM kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P06702
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    14 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human Calprotectin protein
  • Tested applications:
    IHC-P
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [CPT/1028] antibody to Calprotectin for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Calprotectin
Clonality: Monoclonal
Clone: CPT/1028
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human