Chloroacetyl chloride 98%
:79-04-9
:
:C₂H₂Cl₂O
:MFCD00000725
:105 °C (760 mmHg)
:–22 °C
:1.42 g/cm³
:112.94 g/mol
:Ambient
:201-171-6
:6.1
:13,02074
:1752
:GHS06
:GHS08
:GHS05
:GHS09
- Environmentally Preferable:
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4520 to 1.4540 (20°C, 589 nm)
- GC:≥97.5 %
Frequently Bought Together