2,5-Diphenyloxazole, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: PPO, 2,5-Diphenyl-1,3-oxazole
CAS Number:92-71-7
:
Formula:C₁₅H₁₁NO
MDL Number:MFCD00005306
Boiling Pt:360
Melting Pt:70…72 °C
Density:1.094 g/cm³ (99 °C)
Molecular Weight:221.26 g/mol
:M-Clarity™ quality level = MQ200
EINECS:202-181-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together