HYGEN™ Disposable Microfibre Cloths
Reduce cross-contamination with disposable microfibre cloths.
- Removes 99,7% or more of tested viruses and bacteria with water only
- Microfibre (25,2%), Polyester (11,8%) and non-woven Polypropylene (63%) construction
- Help to reduce cross-contamination with a new single-use cloth for each task
- Faciliate colour coding implementation by assigning a coloured cloth to a specific area
- Built-in zig-zag scrubbers assist to effectively remove dirt and debris
- Compatible with common disinfectants Quad, Chlorine Bleach, and Hydrogen Peroxide
The HYGEN™ Disposable Microfibre Cloth makes the most efficient and effective cleaning option for hospital settings and healthcare facilities. Its cleaning capability picks up everything from dirt, dust, and lint to invisible surface bacteria, all while providing the convenience and efficiency of a disposable cleaning solution.
With Rubbermaid Commercial cleaning products, users can fight tough stains and stubborn dust, prioritise infection control, and eliminate surface bacteria with ease.
Frequently Bought Together