Anti-MRP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-0657R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:MRP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Epitope:501-600/1531
- Form:Liquid
- Gene ID:17250
- Antigen synonyms:Abcc1|Mrp1|ATP-binding cassette sub-family C member 1|LTC4 transporter|Multidrug resistance-associated protein 1|Abcc1a|Mdrap|Leukotriene C(4) transporter|MRP|Abcc1b
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:170 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from mouse MRP1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Mediates export of organic anions and drugs from the cytoplasm. Mediates ATP-dependent transport of glutathione and glutathione conjugates, leukotriene C4, estradiol-17-beta-o-glucuronide, methotrexate, antiviral drugs and other xenobiotics. Confers resistance to anticancer drugs. Hydrolyses ATP with low efficiency.
Type: Primary
Antigen: MRP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 501-600/1531
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat