Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-DAXX Ser668 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-12984R-A680
 :  Bioss
Anti-DAXX Ser668 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-12984R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    DAXX Ser668
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    1616
  • Antigen synonyms:
    Death domain associated protein 6|DAP 6|BING 2|MGC126246|BING2|Death associated protein 6|ETS1 associated protein 1|EAP1|Daxx phospho S668|Fas death domain associated protein|DAXX_HUMAN|hDaxx|EAP 1|p-Daxx phospho S668|DAP6|MGC126245
  • Modification:
    Phosphorylation (Ser668)
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    81 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic phosphopeptide derived from human DAXX around the phosphorylation site of Ser668
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 and Fas. Cell death signals are transduced by death domain (DD) containing adapter molecules and members of the ICE/CED3 protease family. A novel DD containing molecule was recently cloned from mouse, human and monkey and designated Daxx. Daxx is a death domain containing important intermediate in the Fas mediated apoptosis. Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N terminal kinase (JNK) pathway. It is widely expressed in fetal and adult human and mouse tissue, indicating its important function in Fas signaling pathways.

Type: Primary
Antigen: DAXX Ser668
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse