- Pk:96 Tests
- Assay Type:Competitive
- Host:
- Primary antibody reactivity:
- Target protein:Vitamin B12
- Description:Vitamin B12 ELISA kit
- Environmentally Preferable:
A competitive ELISA kit for the quantitative measurement of vitamin B12 in biological fluids.
- Easy, convenient and time-saving method
- Detection Range: 0,781 to 50 ng/ml
- Sensitivity: <0,469 ng/ml
- Assay precision: Intra-assay: CV <8%, inter-assay: CV <10%
- No significant cross-reactivity or interference was observed
Vitamin B12 (Vit-B12) is a water-soluble vitamin that is involved in the metabolism of every cell of the human body: it is a cofactor in DNA synthesis, and in both fatty acid and amino acid metabolism. It plays an important role in the synthesis of myelin, and in the maturation of developing red blood cells in the bone marrow. BioVision’s Vitamin B12 ELISA kit is a competitive ELISA assay for the quantitative measurement of Vit-B12 in serum, plasma, and cell culture supernatants. The density of colour is proportional to the amount of Vit-B12 captured from the samples.
Vitamin B12 (Vit-B12) is a water-soluble vitamin that is involved in the metabolism of every cell of the human body: It is a cofactor in DNA synthesis, and in both fatty acid and amino acid metabolism. It plays an important role in the synthesis of myelin, and in the maturation of developing red blood cells in the bone marrow.
Vitamin B12 ELISA kit is a competitive ELISA assay for the quantitative measurement of Vit-B12 in serum, plasma, and cell culture supernatants. The density of colour is proportional to the amount of Vit-B12 captured from the samples.
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.