Order Entry
Slovenia
ContactUsLinkComponent
TapeStation System, 4150
  732-3991
 :  AGILENT
undefined
TapeStation System, 4150
  732-3991
 :  AGILENT
  • Description:
    4150 TapeStation system
  • Environmentally Preferable:
  • Pk:
    1
  • Details:
    Includes software, vortexer, accessories, plastic consumables, user information and I+F, does not include ScreenTape consumables and reagents
Economic, entry-level system for the analysis of up to 16 DNA and RNA samples per run.
  • Established ScreenTape technology guarantees superior ease-of-use of the system, constant costs per sample for up to 16 samples per run
  • Wide range of RNA and DNA ScreenTape applications available including assays for genomic and cell-free DNA
  • Results are obtained quickly in as little as 1 to 2 minutes per sample, or less than 20 minutes for 16 samples
  • Easily switch between DNA and RNA ScreenTape assays for excellent application flexibility, Integrity standards for RNA (RNA integrity number equivalent, RINe) and genomic DNA (DNA Integrity Number, DIN) available
  • Compact footprint saves precious bench space in the laboratory, seamless upgrade to 4200 TapeStation possible by complete assay compatibility
The new agilent 4150 TapeStation system complements the agilent automated electrophoresis platforms with a new, compact, low-throughput instrument. Building on the success of the proven ScreenTape technology, it allows a quick, easy and reliable analysis of nucleic acids for up to 16 samples per run. This affordable, entry-level system is the ideal solution for the quality control (QC) of RNA and DNA samples in next-generation sequencing (NGS), microarray and qPCR workflows.
 : For research use only. Not for use in diagnostic procedures.