Floxacin Amine, Pharmaceutical Analytical Impurities
Catalog # 1A01000.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:172426-88-9
- Description:Floxacin Amine
- Environmentally Preferable:
- Synonym(s):7-amino-1-cyclopropyl-6-fluoro-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
- Description:Floxacin Amine
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.