MPC series couplings, medical grade
Supplier: CPC COLDER
Certificates
About this item
MPC Series couplings add ease of use and security to your most critical fluid handling applications.
- Easy to operate
- BSE/TSE free materials
- Ergonomic thumb latch
- Sterilisable by autoclave, Et0, e-beam or gamma
These couplings provide optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks.
Operating temperature: −40 to 121 °C (for polycarbonate) and −40 to 149 °C (for polysulphone)
Certifications: USP Class VI, ADCF certified and meets biocompatibility requirements.
Specifications
- Catalog No:
- CPCCMPC17002T03
- CPCCMPC17002T03B25
- CPCCMPC17006T39B25
- CPCCMPC17004T03B25
- Max. operating pressure:
- Vacuum...4,1 bar
- Vacuum...4,1 bar
- Vacuum...4,1 bar
- Vacuum...4,1 bar