Order Entry
Slovenia
ContactUsLinkComponent
96-well plate, Bond Elut™ Certify II
Catalog # 551-0435
Supplier:  VARIAN
96-well plate, Bond Elut™ Certify II
Catalog # 551-0435
Supplier:  VARIAN
Specifications
  • Description:
    VersaPlate preassembled 96-well plate, Certify II, 100 mg
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Bond Elut Certify II is an anion exchange/mixed-mode SPE product suitable for extraction of acidic compounds from samples.
  • Ideal for nonpolar and anionic compounds
  • Optimised for acidic drug analysis
  • Bimodal, nonpolar and strong anion exchange
Bond Elut Certify II was developed specifically for the rapid and effective extraction of acidic drugs and metabolites from urine and other biological matrices. Certify II is a mixed-mode cartridge packed with nonpolar C8 and strong anion-exchange (SAX) sorbent.
It has been optimised for acidic drugs, such as 11-nor-delta-9-tetrahydrocannibinol-carboxylic acid, salicylic acid, ibuprofen, acetaminophen, and other compounds that possess both nonpolar and anionic characteristics. For forensic use.
Well plate tubes
Product Information