Polypropylene Muff Covers with Elastic
Supplier: KEYSTONE CLEANROOM PRODUCTS
Certificates
About this item
Keeps cleaned non sterile production hoses protected from external sources of contamination.
- Manufactured and packed in a controlled cleanroom environment
- Reduces labour costs
- Tear and puncture resistant
- Conveniently packaged in 100 per sealed poly bag
These polypropylene muffs are a great choice in non sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover to protect process hoses.
Poly bagged in packs of 100 to help ensure cleanliness up until point of use.
Delivery: Certificates of Conformance sent with each shipment.
Frequently Bought Together