6-Bromo-4-chloro-2-(trifluoromethyl)quinoline 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1701-28-6
:
Formula:C₁₀H₄BrClF₃N
MDL Number:MFCD00153073
Molecular Weight:310.51 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together