DFDNB (1,5-Difluoro-2,4-dinitrobenzene) 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: DFDNB, 1,5-Difluoro-2,4-dinitrobenzene
CAS Number:327-92-4
:
Formula:C₆H₂F₂N₂O₄
MDL Number:MFCD00007052
Boiling Pt:132
Molecular Weight:204.09 g/mol
Melting Pt:72…74 °C
Storage Temperature:Ambient
EINECS:206-324-0
UN:2811
ADR:6.1 II
:GHS06
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together