3-Bromo-5-(trifluoromethyl)picolinic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:959245-76-2
:
Formula:C7H3BrF3NO2
MDL Number:MFCD11036124
Molecular Weight:270 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together