Boc-L-M-Tyrosine 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Boc-m-Tyr-OH, N-Boc-3-hydroxy-L-phenylalanine
CAS Number:90819-30-0
:
Formula:C₁₄H₁₉NO₅
MDL Number:MFCD01632034
Molecular Weight:281.3 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together