3-(3-Hydroxyphenyl)benzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-Cyano-3'-hydroxybiphenyl
CAS Number:154848-43-8
:
Formula:C₁₃H₉NO
MDL Number:MFCD05854256
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together