Ethyl-4-Hydroxy-1-piperidinecarboxylate
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Ethyl 4-hydroxypiperidine-1-carboxylate, 1-Carbethoxy-4-hydroxypiperidine, 4-Hydroxy-1-piperidinecarboxylic Acid Ethyl Ester
CAS Number:65214-82-6
:
Formula:C₈H₁₅NO₃
MDL Number:MFCD00086880
Density:1.12 g/cm³ (25 °C)
Molecular Weight:173.21 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together