Tetrahydro-2H-pyran-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:873397-34-3
:
Formula:C₆H₁₀O₃
MDL Number:MFCD04114349
Boiling Pt:257…258 °C/750mmHg
Density:1.685 g/cm³ (25 °C)
Molecular Weight:130.14 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together