trans-2-Butene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:624-64-6
:
Formula:C₄H₈
Boiling Pt:1 °C
Density:0.6 g/cm³ (25 °C)
Molecular Weight:56.11 g/mol
Melting Pt:–105 °C
Flash Pt:<–30 °C
Storage Temperature:Ambient
EINECS:210-855-3
UN:1012
ADR:2.1
:GHS02
:GHS04
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together