3-Bromo-2-chloro-6-methyl-5-nitropyridine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:856834-95-2
:
Formula:C₆H₄BrClN₂O₂
MDL Number:MFCD09839270
Molecular Weight:251.47 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together