2-Chloro-4-(1-pyrrolidinyl)benzenecarboxylic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2-Chloro-4-pyrrolidinobenzoic Acid, 2-Chloro-4-(pyrrolidin-1-yl)benzoic acid
CAS Number:192513-60-3
:
Formula:C₁₁H₁₂ClNO₂
MDL Number:MFCD03001210
Molecular Weight:225.68 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together