Dimethyl-4-aminothiophene-2,3-dicarboxylate
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Dimethyl 4-aminothiophene-2,3-dicarboxylate
CAS Number:62947-31-3
:
Formula:C₈H₉NO₄S
MDL Number:MFCD00205188
Molecular Weight:215.23 g/mol
Melting Pt:60…61 °C
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together