Order Entry
Slovenia
ContactUsLinkComponent
1,3-Benzothiazole-2-carbaldehyde 98%
1,3-Benzothiazole-2-carbaldehyde 98%
Catalog # APOSOR23088-5G
Supplier:  Apollo Scientific
CAS Number:  
1,3-Benzothiazole-2-carbaldehyde 98%
Catalog # APOSOR23088-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

 :
 :163.2 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:6639-57-2
Formula:C₈H₅NOS
MDL Number:MFCD00526215
Melting Pt:72…76 °C