2-Aminoisonicotinic acid hydrazide
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2-Aminoisonicotinohydrazide, 2-Amino-isonicotinic acid hydrazide
CAS Number:58481-01-9
:
Formula:C₆H₈N₄O
MDL Number:MFCD03791263
Molecular Weight:152.1539 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together