Mal-amido-PEG2-NHS
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Maleimide-PEG2-succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 3-(2-(2-(3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido)ethoxy)ethoxy)propanoate, N-Succinimidyl 13-Maleimido-11-oxo-4,7-dioxa-10-azatridecanoate, 13-Maleimido-11-oxo-4,7-dioxa-10-azatridecanoic acid N-Succinimidyl Ester, Maleimide-PEG2-NHS ester, 2,5-Doxopyrrolidin-1-yl 3-[2-[2-[3-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido]ethoxy]ethoxy]propanoate
CAS Number:955094-26-5
:
Formula:C₁₈H₂₃N₃O₉
MDL Number:MFCD11041137
Molecular Weight:425.39 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together