1,4-Diacryloylpiperazine
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1,1'-(Piperazine-1,4-diyl)bis(prop-2-en-1-one)
CAS Number:6342-17-2
:
Formula:C₁₀H₁₄N₂O₂
MDL Number:MFCD00077661
Molecular Weight:194.23 g/mol
Melting Pt:91.5…93.5 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together