Triacetyllevoglucosan 99% min
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1,6-Anhydro-β-D-glucose-2,3,4-tri-O-acetate
CAS Number:13242-55-2
:
Formula:C₁₂H₁₆O₈
Molecular Weight:288.252 g/mol
Melting Pt:109…110 °C
Storage Temperature:Freezer
EINECS:236-222-1
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together