Sodium chloride solution 2 M, in single-use containers, HyClone™ for bioprocessing
: Saline, NaCl, Sea salt, Halite
:7647-14-5
:
:NaCl
:MFCD00003477
:1413
:58.44 g/mol
:801
:14,08599
:Ambient
- Environmentally Preferable:
- Storage:Room Temperature as defined by regional Pharmacopeia (15-30°C in US; 15-25°C in EU)
- Endotoxin Concentration:≤ 1 EU/mL
- Certifications/Compliance:Manufactured in a ISO9001 and ISO13485 certified facility
- Shelf life:12 Months
- For use with:Chromatography resins, IC systems, ÄKTA hardware systems
- Appearance:Clear, colorless solution
- Sterility:No Growth
Frequently Bought Together