8-(Trifluoromethyl)chroman-4-one 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:890839-66-4
:
Formula:C₁₀H₇F₃O₂
MDL Number:MFCD11506686
Molecular Weight:216.157 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together