1H,1H,2H,2H-Perfluorooctyl bromide
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1-Bromo-1H,1H,2H,2H-perfluorooctane
CAS Number:161583-34-2
:
Formula:C₈H₄BrF₁₃
MDL Number:MFCD03840352
Molecular Weight:427.001 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together